Skip to Content
Merck
All Photos(1)

Key Documents

575069

Sigma-Aldrich

4-Amino-3,5-dimethylisoxazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2O
CAS Number:
Molecular Weight:
112.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 575069, please contact your local Merck office or dealer. Contact Technical Service

Assay

97%

form

solid

mp

41-46 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1noc(C)c1N

InChI

1S/C5H8N2O/c1-3-5(6)4(2)8-7-3/h6H2,1-2H3

InChI key

INSUSOZBMWJGDG-UHFFFAOYSA-N

General description

4-Amino-3,5-dimethylisoxazole can be prepared via reduction of 3,5-dimethyl-4-nitroisoxazole in the presence of Zn/NH4Cl in H2O.[1]

Application

4-Amino-3,5-dimethylisoxazole may be used in the synthesis of 3,5-dimethyl-4-phenylazoisoxazole via reaction with nitrosobenzene.[1]The diazonium salt of 4-Amino-3,5-dimethylisoxazole can react with cupric sulfate to form acetyltriazole derivatives.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grunanger P and Vita-Finzi P
Isoxazoles, Volume 49, part 1 (1991)
"Transformations in the Isoxazole Series: Synthesis of Substituted 2-Aminothiazoles"
Pascual A, et al.
Helvetica Chimica Acta, 74(3), 531-542 (1991)
Transformations in the Isoxazole Series: Synthesis of Substituted 2-Aminothiazoles
Pascual A, et al.
Helvetica Chimica Acta, 74(3), 531-542 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service