Skip to Content
Merck
All Photos(1)

Key Documents

56420

Sigma-Aldrich

4(3H)-Pyrimidinone

≥98.0% (NT)

Synonym(s):

4(3H)-Pyrimidone, 4-Hydroxypyrimidine, 4-Pyrimidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O
CAS Number:
Molecular Weight:
96.09
Beilstein:
609975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

solid

mp

163-167 °C
166-169 °C (lit.)

SMILES string

O=C1C=CNC=N1

InChI

1S/C4H4N2O/c7-4-1-2-5-3-6-4/h1-3H,(H,5,6,7)

InChI key

DNCYBUMDUBHIJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Harris et al.
The Journal of biological chemistry, 272(36), 22514-22525 (1997-09-05)
Isolated from bovine milk, xanthine oxidase (XO) and xanthine dehydrogenase (XDH) are two interconvertible forms of the same protein, differing in the number of protein cysteines versus cystines. Most differences between XO and XDH are localized to the FAD center
Raquel Sanchez et al.
Journal of the American Chemical Society, 129(19), 6287-6290 (2007-04-24)
4-Hydroxypyrimidine (4HP) has two conformational forms (the hydroxyl hydrogen cis or trans with respect to the adjacent nitrogen), which are in tautomeric equilibrium with two ketonic forms, 4-pyrimidinone (4PO) and 6-pyrimidinone (6PO). We have investigated the free jet absorption millimeterwave
Aleem Gangjee et al.
Bioorganic & medicinal chemistry, 18(10), 3575-3587 (2010-04-21)
A series of eight N(4)-phenylsubstituted-6-(2,4-dichlorophenylmethyl)-7H-pyrrolo[2,3-d]pyrimidine-2,4-diamines 8-15 were synthesized as vascular endothelial growth factor receptor-2 (VEGFR-2) inhibitors with varied substitutions in the phenyl ring of the 4-anilino moiety. In addition, five N(4)-phenylsubstituted-6-phenylmethylsubstituted-7H-pyrrolo[2,3-d]pyrimidin-4-amines 16-20 were synthesized to evaluate the importance of the
Barbara M Giuliano et al.
The journal of physical chemistry. A, 114(48), 12725-12730 (2010-11-10)
The keto-enol tautomerism of 4-hydroxypyrimidine and of the related molecules S-methyl-2-thiouracil and 2-thiouracil has been investigated using synchrotron-based techniques. The populations of the constituent tautomers and thermodynamic parameters have been obtained by analysis of core-level photoemission spectra. The effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service