545155
1-Butenyl ethyl ether, mixture of cis and trans
97%
Synonym(s):
1-Ethoxy-1-butene
About This Item
Recommended Products
Assay
97%
refractive index
n20/D 1.4060
bp
94-95 °C (lit.)
density
0.778 g/mL at 25 °C (lit.)
SMILES string
CCO\C=C\CC
InChI
1S/C6H12O/c1-3-5-6-7-4-2/h5-6H,3-4H2,1-2H3/b6-5+
InChI key
AQTYNINXYJFSHD-AATRIKPKSA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
44.0 °F - closed cup
Flash Point(C)
6.67 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service