Skip to Content
Merck
All Photos(1)

Documents

535141

Sigma-Aldrich

Poly(isobutylene-co-maleic acid) sodium salt

crosslinked, powder, <900 μm particle size (95%)

Synonym(s):

Isobutylene-maleic anhydride copolymer sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C4H8)m(C4H4O4)n · xNa
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

crosslinked, powder

particle size

<900 μm (95%)

capacity

200 mL/g absorption capacity (distilled water)
30 mL/g, absorption (4 wt. % saline solution)

mp

>300 °C

density

1.3 g/mL at 25 °C (lit.)

SMILES string

CC(C)=C.OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(=O)O[Na]

Looking for similar products? Visit Product Comparison Guide

General description

Superabsorbent polymer. Minimal water released under pressure. Good stability in hot and cold water. Moisture released at lower relative humidity. Stable up to ~280°C

Application

Used in hygiene products to increase liquid absorbency, as soil additive to increase water retaining capacity, as dehydrating agent in construction applications, and coagulating agent for mud to prevent outflow of mud.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service