Skip to Content
Merck
All Photos(1)

Key Documents

534625

Sigma-Aldrich

4-Carboxybenzenesulfonamide, polymer-bound

50-90 mesh, extent of labeling: 0.8-1.0 mmol/g loading, 1 % cross-linked

Synonym(s):

4-Sulfamylbenzoyl resin, Kenner’s "Safety-Catch"resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

0.8-1.0 mmol/g loading

particle size

50-90 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.NS(=O)(=O)c3ccc(cc3)C(=O)NCc4ccccc4

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service