Skip to Content
Merck
All Photos(1)

Key Documents

528234

Sigma-Aldrich

3-Ethoxybenzyl alcohol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CH2OH
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.534 (lit.)

bp

266 °C (lit.)

density

1.08 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCOc1cccc(CO)c1

InChI

1S/C9H12O2/c1-2-11-9-5-3-4-8(6-9)7-10/h3-6,10H,2,7H2,1H3

InChI key

PDYCXUZHSKLETQ-UHFFFAOYSA-N

General description

3-Ethoxybenzyl alcohol can be obtained from m-anisaldehyde.[1] It undergoes reduction in the presence of LiAlH4 and ether to afford 3-ethoxybenzyl alcohol.[2]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ward OP.
Bioprocessing, 161-161 (2012)
Flowers JL, et al.
Cracking the MCAT with CD-ROM, 868-868 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service