Skip to Content
Merck
All Photos(1)

Key Documents

527327

Sigma-Aldrich

PSS-Trivinylsilyloxy-Heptacyclopentyl substituted

Synonym(s):

1-(Trivinylsilyloxy)-3,5,7,9,11,13,15-heptacyclopentylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Trivinylsilyloxy-POSS®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H72O13Si9
CAS Number:
Molecular Weight:
1025.77
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

>350 °C (lit.)

SMILES string

C=C[Si](O[Si]12O[Si]3(O[Si]4(O[Si](O1)(O[Si]5(O[Si](O2)(O[Si](O3)(O[Si](O4)(O5)C6CCCC6)C7CCCC7)C8CCCC8)C9CCCC9)C%10CCCC%10)C%11CCCC%11)C%12CCCC%12)(C=C)C=C

InChI

1S/C41H72O13Si9/c1-4-55(5-2,6-3)42-63-52-60(39-29-15-16-30-39)46-57(36-23-9-10-24-36)43-56(35-21-7-8-22-35)44-58(48-60,37-25-11-12-26-37)50-62(54-63,41-33-19-20-34-41)51-59(45-56,38-27-13-14-28-38)49-61(47-57,53-63)40-31-17-18-32-40/h4-6,35-41H,1-3,7-34H2

InChI key

YXPZADQYIFWKNL-UHFFFAOYSA-N

Legal Information

POSS is a registered trademark of Hybrid Plastics, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service