Skip to Content
Merck
All Photos(1)

Key Documents

517526

Sigma-Aldrich

Scandium(III) bromide

anhydrous, powder, ≥99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ScBr3
CAS Number:
Molecular Weight:
284.67
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: scandium

SMILES string

Br[Sc](Br)Br

InChI

1S/3BrH.Sc/h3*1H;/q;;;+3/p-3

InChI key

APPHYFNIXVIIJR-UHFFFAOYSA-K

Application

Employed in solid state synthesis of unusual clusters, Sc19Br28Z4, (Z=Mn, Fe, Ru or Os). These clusters are of interest for their structure and magnetic properties.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steinwand, S.J. et al.
Inorganic Chemistry, 36, 6413-6413 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service