Recommended Products
Assay
98%
refractive index
n20/D 1.575 (lit.)
bp
98-100 °C/25 mmHg (lit.)
density
1.55 g/mL at 25 °C (lit.)
SMILES string
Cc1cc(Br)ccc1Cl
InChI
1S/C7H6BrCl/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3
InChI key
OZFQMHJKAODEON-UHFFFAOYSA-N
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thermophysical Properties of Chemicals and Hydrocarbons, 536-536 (2014)
2, 3-Dichloroindone as a dienophilic compound.
Journal of the American Chemical Society, 81(21), 5641-5646 (1959)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service