Skip to Content
Merck
All Photos(1)

Key Documents

511668

Sigma-Aldrich

1,2-Dichloroethane

anhydrous, 99.8%

Synonym(s):

Ethylene chloride, Ethylene dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2Cl
CAS Number:
Molecular Weight:
98.96
Beilstein:
605264
EC Number:
MDL number:
UNSPSC Code:
12190000
Pricing and availability is not currently available.

grade

anhydrous

vapor density

3.4 (20 °C, vs air)

vapor pressure

87 mmHg ( 25 °C)

Assay

99.8%

autoignition temp.

775 °F

expl. lim.

15.6 %

refractive index

n20/D 1.444 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

ClCCCl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2

InChI key

WSLDOOZREJYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethane (1,2-DCA, EDC) is a halogenated aliphatic hydrocarbon commonly used as a precursor for generating vinyl chloride. Genotoxic studies of EDC have been reported.[1] Structural investigation of EDC by electron diffraction shows that trans-form predominated over gauche-form.[2] The degradation of 1,2-DCA with simultaneous current generation by anodophilic consortia present in microbial fuel cells (MFCs) has been studied.[3] The determination of 1,2-DCA in aqueous solution by an enzyme-based fiber optic biosensor has been reported.[4] The spin-lattice relaxation time of 1,2-DCA in both solid and liquid state has been measured.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review of the genotoxicity of 1,2-dichloroethane (EDC).
Gwinn MR, et al.
Mutation Research, Reviews in Mutation Research, 727(1), 42-53 (2011)
Development of a fiber optic enzymatic biosensor for 1, 2-dichloroethane.
Campbell DW, et al.
Biotechnology Letters, 28(12), 883-887 (2006)
Nuclear Magnetic Relaxation in 1,2-Dichloroethane.
Linder S.
J. Chem. Phys. , 26(4), 900-905 (1957)
The Structure and Internal Motion of 1,2-Dichloroethane.
Ainsworth J and Karle J.
J. Chem. Phys. , 20(3), 425-427 (1952)
Enhanced removal of 1,2-dichloroethane by anodophilic microbial consortia.
Pham H, et al.
Water Research, 43(11), 2936-2946 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service