Skip to Content
Merck
All Photos(1)

Documents

50620

Sigma-Aldrich

Butyl glycolate

technical, ≥90% (GC)

Synonym(s):

Butyl hydroxyacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2COO(CH2)3CH3
CAS Number:
Molecular Weight:
132.16
Beilstein:
1749477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.427

bp

187-190 °C (lit.)

density

1.019 g/mL at 20 °C (lit.)

SMILES string

CCCCOC(=O)CO

InChI

1S/C6H12O3/c1-2-3-4-9-6(8)5-7/h7H,2-5H2,1H3

InChI key

VFGRALUHHHDIQI-UHFFFAOYSA-N

General description

Butyl glycolate is a glycolic acid derivative.

Application

Butyl glycolate may be used in the preparation of N-(α-hydroxyacetyl)-(S)-isoleucine.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of new block copolymers with poly(ethylene oxide) and poly[3(S)-sec-butylmorpholine-2,5-dione] sequences.
Feng Y, et al.
Macromolecular Bioscience, 1(1), 30-39 (2001)
Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 52, 321-331 (2018-07-22)
The purpose of present study was to investigate the applicability of reconstructed human epidermis model to identify skin corrosive UN GHS Categories 1A, 1B/1C and non-corrosive chemicals in China. By using a commercialized reconstructed human epidermis model, China EpiSkin™ which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service