494305
Vinyltrimethylsilane
≥99.5%
Synonym(s):
Trimethyl(vinyl)silane, (Trimethylsilyl)ethylene, Ethenyltrimethylsilane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3SiCHCH2
CAS Number:
Molecular Weight:
100.23
Beilstein:
956572
EC Number:
MDL number:
UNSPSC Code:
12352300
vapor pressure
4.44 psi ( 20 °C)
Assay
≥99.5%
refractive index
n20/D 1.391 (lit.)
bp
55 °C (lit.)
density
0.684 g/mL at 25 °C (lit.)
SMILES string
C[Si](C)(C)C=C
InChI
1S/C5H12Si/c1-5-6(2,3)4/h5H,1H2,2-4H3
InChI key
GCSJLQSCSDMKTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-11.2 °F
Flash Point(C)
-24 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service