Skip to Content
Merck
All Photos(1)

Documents

484660

Sigma-Aldrich

4-Cyanophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4NCS
CAS Number:
Molecular Weight:
160.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

119-123 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=Nc1ccc(cc1)C#N

InChI

1S/C8H4N2S/c9-5-7-1-3-8(4-2-7)10-6-11/h1-4H

InChI key

DZFKAXLNKZXNHD-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Feng Cheng et al.
Chemistry, an Asian journal, 12(1), 45-51 (2016-10-27)
Herein, two molecules based on urea and thiourea, which differ by only a single atom, were designed, successfully synthesized, and fabricated into resistive random-access memory devices (RRAM). The urea-based molecule showed binary write-once-read-many (WORM) storage behavior, whereas the thiourea-based molecule
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service