Skip to Content
Merck
All Photos(2)

Key Documents

480185

Sigma-Aldrich

2-Chloro-5-chlorosulfonyl-4-fluorobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(SO2Cl)(F)CO2H
CAS Number:
Molecular Weight:
273.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

145-148 °C (lit.)

SMILES string

OC(=O)c1cc(c(F)cc1Cl)S(Cl)(=O)=O

InChI

1S/C7H3Cl2FO4S/c8-4-2-5(10)6(15(9,13)14)1-3(4)7(11)12/h1-2H,(H,11,12)

InChI key

QOPIIRYHSQOQRN-UHFFFAOYSA-N

General description

2-Chloro-5-chlorosulfonyl-4-fluorobenzoic acid is an aryl fluorinated building block used in organic synthesis. It participates in the synthesis of N-alkylated 4-substituted methylsulfonylanthranilic acid derivatives.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aminobenzoic acid diuretics. 8. 3,4-Disubstituted 5-methylsulfonylbenzoic acids and related compounds.
Feit PW and Nielsen OBT.
Journal of Medicinal Chemistry, 19(3), 402-406 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service