Skip to Content
Merck
All Photos(1)

Key Documents

474185

Sigma-Aldrich

Glycidyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononyl ether

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8F16O2
CAS Number:
Molecular Weight:
488.17
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

96%

refractive index

n20/D 1.342 (lit.)

bp

251 °C (lit.)

density

1.682 g/mL at 25 °C (lit.)

SMILES string

FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1

InChI

1S/C12H8F16O2/c13-5(14)7(17,18)9(21,22)11(25,26)12(27,28)10(23,24)8(19,20)6(15,16)3-29-1-4-2-30-4/h4-5H,1-3H2

InChI key

AVKVPTNDWAGIMH-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Lact. - Repr. 1B - STOT RE 1

Target Organs

Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service