Skip to Content
Merck
All Photos(1)

Key Documents

465062

Sigma-Aldrich

Benzyl (S)-(−)-1,2,3,4-tetrahydro-3-isoquinolinecarboxylate p-toluenesulfonic acid salt

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO2 · C7H8O3S
CAS Number:
Molecular Weight:
439.52
Beilstein:
5722289
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

145-150 °C (lit.)

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.O=C(OCc2ccccc2)[C@@H]3Cc4ccccc4CN3

InChI

1S/C17H17NO2.C7H8O3S/c19-17(20-12-13-6-2-1-3-7-13)16-10-14-8-4-5-9-15(14)11-18-16;1-6-2-4-7(5-3-6)11(8,9)10/h1-9,16,18H,10-12H2;2-5H,1H3,(H,8,9,10)/t16-;/m0./s1

InChI key

PSMBIFNNFMRIMV-NTISSMGPSA-N

Application

Used to prepare a novel class of antifungal agents.[1]

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu, L.T. et al.
Bioorganic & Medicinal Chemistry Letters, 6, 1335-1335 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service