Skip to Content
Merck
All Photos(1)

Documents

457035

Sigma-Aldrich

4-(Trifluoromethyl)hydrocinnamic acid

95%

Synonym(s):

3-[4-(Trifluoromethyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
218.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

106-110 °C (lit.)

SMILES string

OC(=O)CCc1ccc(cc1)C(F)(F)F

InChI

1S/C10H9F3O2/c11-10(12,13)8-4-1-7(2-5-8)3-6-9(14)15/h1-2,4-5H,3,6H2,(H,14,15)

InChI key

OEIUMLSCWINLBB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-[4-(Trifluoromethyl) phenyl] propanoic acid.
Guan J-N, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(4), o844-o844 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service