Skip to Content
Merck
All Photos(1)

Key Documents

451371

Sigma-Aldrich

Yttrium(III) fluoride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Yttrium fluoride, Yttrium trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YF3
CAS Number:
Molecular Weight:
145.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: yttrium

density

4.01 g/mL at 25 °C (lit.)

SMILES string

F[Y](F)F

InChI

1S/3FH.Y/h3*1H;/q;;;+3/p-3

InChI key

RBORBHYCVONNJH-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Recent applications include studies of InF3-based glasses and optical characterization of YF3 thin films.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanko Nguyen et al.
Colloids and surfaces. B, Biointerfaces, 136, 744-751 (2015-11-01)
The in vitro adsorption and retention of liposomes onto four common types of dental restorative materials (conventional and silorane-based resin composites as well as conventional and resin-modified glass ionomer cements (GIC)) have been investigated due to their potential use in
Journal of Non-Crystalline Solids, 204, 260-260 (1996)
Proc. SPIE: Int. Soc. Opt. Eng., 2776, 366-366 (1996)
Jung-Hoon Ro et al.
Lasers in medical science, 30(5), 1497-1503 (2015-04-22)
The selection of a light-curing unit for the curing composite resins is important to achieve best outcomes. The purpose of the present study was to test lasers of 457 and 473 nm alone or in combination under different light conditions with
Kan Usuda et al.
Biological trace element research, 167(1), 84-90 (2015-03-10)
Various fluoride compounds are widely used in industry. The present risk assessment study was conducted using a series of inorganic binary fluorides of the type XFn, where X(n) = Na(+), K(+), Li(+), Mg(2+), Ca(2+), Sr(2+), Ba(2+), Al(3+), Nd(3+), La(3+), Ce(3+), Sm(3+), Gd(3+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service