All Photos(1)
About This Item
Linear Formula:
F3C6H2CHO
CAS Number:
Molecular Weight:
160.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
96%
refractive index
n20/D 1.487 (lit.)
bp
159 °C (lit.)
density
1.425 g/mL at 25 °C (lit.)
SMILES string
Fc1ccc(F)c(C=O)c1F
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
168.8 °F
Flash Point(C)
76 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service