Skip to Content
Merck
All Photos(1)

Documents

430048

Sigma-Aldrich

Coconut oil from Cocos nucifera

Synonym(s):

Coconut fat, Copra oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211

General description

Coconut oil from Cocos nucifera is the most common vegetable oil. It is mainly composed of lauric acid, which is responsible for most of the physiological and metabolic properties of coconut oil. It also contains high amount of low molecular weight saturated fatty acids, which makes it applicable for both edible and non-edible purposes. Deacidification by ultrafiltration of coconut oil has been reported by membrane filtration. Its decomposition thermally and by oxidation have been analyzed by thermo-gravimetric analysis. Its potential as a source to generate biodiesel has been studied under various operating conditions.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deacidification of coconut oil by membrane filtration.
Rao YPC, et al.
Food and Bioprocess Technology, 6(2), 498-508 (2013)
The properties of Lauric acid and their significance in coconut oil.
Dayrit FM.
Journal of the American Oil Chemists' Society, 92(1), 1-15 (2015)
Enzymatic conversion of coconut oil for biodiesel production.
Tupufia SC, et al.
Fuel Processing Technology, 106, 721-726 (2013)
Coconut oil as base oil for industrial lubricants-evaluation and modification of thermal, oxidative and low temperature properties.
Jayadas NH and Prabhakaran Nair K.
Tribology International, 39(9), 873-878 (2006)
Chemical properties of virgin coconut oil.
Marina AM, et al.
Journal of the American Oil Chemists' Society, 86(4), 301-307 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service