Skip to Content
Merck
All Photos(1)

Key Documents

428949

Sigma-Aldrich

4-Methyl-3-nitrobenzoyl chloride

99%

Synonym(s):

3-Nitro-p-toluoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)COCl
CAS Number:
Molecular Weight:
199.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.581 (lit.)

bp

185 °C/36 mmHg (lit.)

mp

20-21 °C (lit.)

density

1.37 g/mL at 25 °C (lit.)

functional group

acyl chloride
nitro

SMILES string

Cc1ccc(cc1[N+]([O-])=O)C(Cl)=O

InChI

1S/C8H6ClNO3/c1-5-2-3-6(8(9)11)4-7(5)10(12)13/h2-4H,1H3

InChI key

DXMHBBURYDVYAI-UHFFFAOYSA-N

General description

4-Methyl-3-nitrobenzoyl chloride is a benzoyl chloride derivative. It has been reported to be synthesized from 4-methyl-3-nitrobenzoic acid.[1]

Application

4-Methyl-3-nitrobenzoyl chloride was used in the synthesis of 4-amino-1,5-naphthalenedisulphonate acid monosodium salt, an intermediate employed in the synthesis of modified suramin molecule.[2]
It may be used in the synthesis of the following:
  • benzophenone derivative[3]
  • substituted 3-amino-4-methyl-N-phenylbenzamide[4]
  • retroamide[5]
  • 4-methyl-3-nitro-N-phenylbenzamide[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M J Ashton et al.
Journal of medicinal chemistry, 39(17), 3343-3356 (1996-08-16)
The synthesis and biological activity of a new series of benzamides and related compounds that upregulate the expression of the low-density lipoprotein (LDL) receptor in human hepatocytes (HepG2 cells) by a novel mechanism are described. The lead compound, N-[5-[(3-cyclohexylpropionyl)amino]-2-methylphenyl]-4-hydroxybe nzamide
Holly L Deak et al.
Bioorganic & medicinal chemistry letters, 18(3), 1172-1176 (2007-12-18)
N-3-(Phenylcarbamoyl)arylpyrimidine-5-carboxamides are a novel class of selective Lck inhibitors. This series of compounds derives its selectivity from a hydrogen bond with the gatekeeper Thr316 of the enzyme. X-ray co-crystal structural data, structure-activity relationships, and the synthesis of these inhibitors are
Shuxin Li et al.
Bioorganic & medicinal chemistry letters, 22(16), 5279-5282 (2012-07-14)
A series of acrylamide analogues were designed and synthesized from Imatinib and Nilotinib as novel BCR-ABL inhibitors by application of the principle of nonclassical electronic isostere. All new compounds were evaluated for their inhibitory effects on the activity of BCR-ABL
Romina Croci et al.
PloS one, 9(3), e91765-e91765 (2014-03-14)
Noroviruses (NV) are +ssRNA viruses responsible for severe gastroenteritis; no effective vaccines/antivirals are currently available. We previously identified Suramin (9) as a potent inhibitor of NV-RNA dependent RNA polymerase (NV-RdRp). Despite significant in vitro activities versus several pharmacological targets, Suramin
Marie-Pierre Lézé et al.
Bioorganic & medicinal chemistry letters, 18(16), 4713-4715 (2008-07-22)
Two new series of benzonitrile derivatives on position 6 or 4 of indole ring were successfully synthesized via a Leimgruber-Batcho reaction. All the compounds were evaluated in vitro on the inhibition of aromatase (CYP19) and 17alpha-hydroxylase-C17,20-lyase (CYP17). The racemate, 4-[(1H-imidazol-1-yl)(1H-indol-4-yl)methyl]benzonitrile

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service