Skip to Content
Merck
All Photos(1)

Key Documents

422258

Sigma-Aldrich

3-(Trifluoromethyl)benzamidoxime

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4C(=NOH)NH2
CAS Number:
Molecular Weight:
204.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

87-90 °C (lit.)

SMILES string

N\C(=N\O)c1cccc(c1)C(F)(F)F

InChI

1S/C8H7F3N2O/c9-8(10,11)6-3-1-2-5(4-6)7(12)13-14/h1-4,14H,(H2,12,13)

InChI key

SBGBSARAGZEWGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-(Trifluoromethyl)benzamidoxime is a benzamidoxime (BAO) derivative containing amidoxime functional group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takayuki Shibata et al.
Analytica chimica acta, 674(2), 234-238 (2010-08-04)
Facile and specific methods to quantify a nucleobase in biological samples are of great importance for diagnosing disorders in nucleic acid metabolism. In the present study, a novel fluorogenic reaction specific for uracil has been developed. The reaction was carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service