Skip to Content
Merck
All Photos(2)

Documents

419664

Sigma-Aldrich

Allyl 2,4,6-tribromophenyl ether

98%

Synonym(s):

1,3,5-Tribromo-2-(2-propen-1-yloxy)benzene, 2,4,6-Tribromophenyl allyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br3C6H2OCH2CH=CH2
CAS Number:
Molecular Weight:
370.86
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

mp

74-76 °C (lit.)

SMILES string

Brc1cc(Br)c(OCC=C)c(Br)c1

InChI

1S/C9H7Br3O/c1-2-3-13-9-7(11)4-6(10)5-8(9)12/h2,4-5H,1,3H2

InChI key

RZLLIOPGUFOWOD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinjing Ma et al.
Environmental pollution (Barking, Essex : 1987), 267, 115497-115497 (2020-09-06)
The study aimed to investigate temporal trends of "old" and "new" persistent halogenated organic pollutants (HOPs) in Taihu Lake, the third largest freshwater lake in China, and the associated health risks. Five fish species were consecutively collected from the lake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service