Skip to Content
Merck
All Photos(1)

Key Documents

410551

Sigma-Aldrich

[2,6-Dichloro-4-(trifluoromethyl)phenyl]hydrazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(CF3)NHNH2
CAS Number:
Molecular Weight:
245.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

58-60 °C (lit.)

SMILES string

NNc1c(Cl)cc(cc1Cl)C(F)(F)F

InChI

1S/C7H5Cl2F3N2/c8-4-1-3(7(10,11)12)2-5(9)6(4)14-13/h1-2,14H,13H2

InChI key

FYOWOHMZNWQLFG-UHFFFAOYSA-N

General description

[2,6-Dichloro-4-(trifluoromethyl)phenyl]hydrazine is a hydrazine derivative.[1] It reacts with excess of ethyl acetoacetate to afford 1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-3,4-dimethylpyrano[2,3-c]pyrazol-6(1H)-one.[2]

Application

[2,6-Dichloro-4-(trifluoromethyl)phenyl]hydrazine may be used for the synthesis of pyrazolo[4,3-d]isoxazole derivatives.[1] It may also be used in the synthesis of following N-(2,6-dichloro-4-trifluoromethyl)phenyl-N′-(1-phenylethylidene) hydrazines:[3]
  • N-(2,6-dichloro-4-trifluoromethyl)phenyl-N′-(1-phenylethylidene)hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(4-chlorophenyl)ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(3-chlorophenyl)ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(4-bromophenyl)-ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(3-bromophenyl)ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(4-methoxyphenyl)ethylidene]-hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(4-trifluoromethylphenyl)-ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(4-nitrophenyl)ethylidene]hydrazine
  • N-(2,6-Dichloro-4-trifluoromethyl)phenyl-N′-[1-(6-methoxy-naphthalen-2-yl)-ethylidene]hydrazine

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SYNTHESIS AND ANTIBACTERIAL ACTIVITY OF FUSED ISOXAZOLE DERIVATIVES USING GRINDING METHOD.
Badrey MG and Gomha SM.
International Journal of Pharmacy and Pharmaceutical Sciences, 6(7) (2014)
Huanan Hu et al.
Molecules (Basel, Switzerland), 15(10), 7472-7481 (2010-10-27)
A series of novel 1-[(2,6-dichloro-4-trifluoromethyl)phenyl]-3-aryl-¹H-pyrazole-4-carbaldehydes were synthesized using the Vilsmeier-Haack reagent. The structures of all the title compounds have been confirmed by elemental analysis, ¹H-NMR and ¹³C-NMR and in addition, the structure of intermediate 5b was investigated by X-ray crystallography.
Hoong-Kun Fun et al.
Acta crystallographica. Section E, Structure reports online, 68(Pt 8), o2350-o2350 (2012-08-21)
In the title compound, C(15)H(9)Cl(2)F(3)N(2)O(2), the 1,6-dihydro-pyrano[2,3-c]pyrazole ring system is almost planar, with a maximum deviation of 0.0226 (14) Å, and forms a dihedral angle of 69.90 (6)° with the benzene ring. In the crystal, mol-ecules are linked into a helical chain along

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service