Skip to Content
Merck
All Photos(2)

Documents

400424

Sigma-Aldrich

6-Hydroxy-2-naphthalenesulfonic acid sodium salt hydrate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC10H6SO3Na · xH2O
Molecular Weight:
246.21 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

form

solid

mp

>300 °C (lit.)

SMILES string

O.[Na+].Oc1ccc2cc(ccc2c1)S([O-])(=O)=O

InChI

1S/C10H8O4S.Na.H2O/c11-9-3-1-8-6-10(15(12,13)14)4-2-7(8)5-9;;/h1-6,11H,(H,12,13,14);;1H2/q;+1;/p-1

InChI key

QHVRFNCHOMKXQP-UHFFFAOYSA-M

General description

Sodium salt of 6-hydroxy-2-naphthalenesulfonic acid is also referred as Schaeffer′s salt.

Application

6-Hydroxy-2-naphthalenesulfonic acid sodium salt hydrate may be used in the following studies:
  • As precursors during the agar-plate screening test.
  • Preparation of methoxy-substituted naphthalenesulfonyl chloride.
  • Synthesis of FD&C Red No. 40, a synthetic water-soluble color that is permitted for coloring foods, drugs and cosmetics in the U.S.A.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anamarija Zega et al.
Bioorganic & medicinal chemistry letters, 14(6), 1563-1567 (2004-03-10)
A series of azaphenylalanine derivatives were investigated as novel thrombin inhibitors based on the prodrug principle. By systematic structural modifications we have identified optimal groups for this series that led us to potent inhibitors of thrombin incorporating the benzamidine fragment
N Richfield-Fratz et al.
Journal of chromatography, 467(1), 167-176 (1989-04-21)
The unsulfonated aromatic amine 4-nitro-p-cresidine (2-methoxy-5-methyl-4-nitrobenzenamine) was identified as an impurity in the regulated color additive FD&C Red. No. 40. The compound was isolated from the water-soluble color by extraction with chloroform, followed by transfer of the free amines to
Jolanta Polak et al.
Microbial cell factories, 9, 51-51 (2010-07-06)
Chemical methods of producing dyes involve extreme temperatures and unsafe toxic compounds. Application of oxidizing enzymes obtained from fungal species, for example laccase, is an alternative to chemical synthesis of dyes. Laccase can be replaced by fungal biomass acting as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service