Skip to Content
Merck
All Photos(1)

Key Documents

398454

Sigma-Aldrich

Potassium tetrabromoaurate(III) hydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KAuBr4 2H2O
CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
solid

Quality Level

Assay

99.9% trace metals basis

form

solid

impurities

≤1500.0 ppm Trace Metal Analysis

SMILES string

[Br-][Au+3]([Br-])([Br-])[Br-].[K+].O.O

InChI

1S/Au.4BrH.K.H2O/h;4*1H;;1H2/q+3;;;;;+1;/p-4

InChI key

KKQNBALBBILMEX-UHFFFAOYSA-J

Application

An aqueous solution of KAuBr4 can be used for galvanic replacement of the liquid metal galinstan.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Abu Sayeed et al.
Chemistry, an Asian journal, 15(24), 4339-4346 (2020-11-06)
Although the presence of iron in mixed metal oxide based catalysts has shown significant performance improvement in the oxygen evolution reaction (OER), iron oxides themselves demonstrate much poorer activity. In this study, we investigate improving the performance of iron catalysts
Galvanic Replacement of the Liquid Metal Galinstan.
Hoshyargar F, et al.
Journal of the American Chemical Society, 139(4), 1464-1471 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service