Skip to Content
Merck
All Photos(3)

Documents

394009

Sigma-Aldrich

(R)-(+)-N,α-Dimethylbenzylamine

98%

Synonym(s):

(R)-(+)-N-Methyl-1-phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NHCH3
CAS Number:
Molecular Weight:
135.21
Beilstein:
4350160
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]20/D +70°, c = 2 in chloroform

refractive index

n20/D 1.511 (lit.)

bp

74-76 °C/11 mmHg (lit.)

density

0.924 g/mL at 20 °C (lit.)

SMILES string

CN[C@H](C)c1ccccc1

InChI

1S/C9H13N/c1-8(10-2)9-6-4-3-5-7-9/h3-8,10H,1-2H3/t8-/m1/s1

InChI key

RCSSHZGQHHEHPZ-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to make asymmetric organometallic catalysts (e.g., for conjugate additions to enones).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 31, 965-965 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service