Skip to Content
Merck
All Photos(1)

Key Documents

39277

Sigma-Aldrich

(3S,6S)-2,7-Dimethyl-3,6-octanediol

≥99.0% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H22O2
CAS Number:
Molecular Weight:
174.28
Beilstein:
4738501
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

Assay

≥99.0% (sum of enantiomers, GC)

optical purity

enantiomeric ratio: ≥99:1

SMILES string

CC(C)[C@@H](O)CC[C@H](O)C(C)C

InChI

1S/C10H22O2/c1-7(2)9(11)5-6-10(12)8(3)4/h7-12H,5-6H2,1-4H3/t9-,10-/m0/s1

InChI key

IIEGQVRKIRPFFP-UWVGGRQHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service