389323
Poly(vinyl chloride)
average Mw ~80,000, average Mn ~47,000
Synonym(s):
PVC
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
powder
mol wt
average Mn ~47,000
average Mw ~80,000
inherent viscosity
0.80 dL/g(lit.)
density
1.4 g/mL at 25 °C (lit.)
application(s)
battery manufacturing
SMILES string
ClC=C
InChI
1S/C2H3Cl/c1-2-3/h2H,1H2
InChI key
BZHJMEDXRYGGRV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
British journal of anaesthesia, 110(2), 305-310 (2012-11-21)
In a previous study, the authors found a large bias (50%) for lithium (LiDCO) compared with thermodilution cardiac output measurement methods in ponies receiving i.v. infusions of xylazine, ketamine, and midazolam. This prompted the authors to examine the effect of
Applied spectroscopy, 66(11), 1286-1293 (2012-11-14)
Airborne silica dust (quartz) is common in coal mines and represents a respiratory hazard that can lead to silicosis, a potentially fatal lung disease. With an eye toward developing a portable monitoring device for rapid analysis of silica dust, laser-induced
Environmental science & technology, 46(21), 11566-11574 (2012-10-11)
Opportunistic pathogens represent a unique challenge because they establish and grow within drinking water systems, yet the factors stimulating their proliferation are largely unknown. The purpose of this study was to examine the influence of pipe materials, disinfectant type, and
Critical care medicine, 41(2), 518-526 (2012-12-25)
To assess the structural characteristics involved in the design of high-volume low-pressure endotracheal tube cuffs that are associated with fluid sealing effectiveness and to determine the extent of transmitted tracheal pressures upon cuff inflation. In vitro study. Pneumology laboratories. Eight
International journal of food microbiology, 159(3), 179-185 (2012-10-31)
The aim of this study was to determine how quickly the surface of a refrigerated supermarket serve over counter becomes loaded with bacteria. New material made of polyvinyl chloride or stainless steel was placed on the surface on which foodstuffs
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service