Skip to Content
Merck
All Photos(1)

Documents

374962

Sigma-Aldrich

4-Guanidinobenzoic acid hydrochloride

99%

Synonym(s):

4-Guanidobenzoic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC(=NH)NHC6H4CO2H · HCl
CAS Number:
Molecular Weight:
215.64
Beilstein:
3718032
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

285 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].NC(=N)Nc1ccc(cc1)C(O)=O

InChI

1S/C8H9N3O2.ClH/c9-8(10)11-6-3-1-5(2-4-6)7(12)13;/h1-4H,(H,12,13)(H4,9,10,11);1H

InChI key

YETFLAUJROGBMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Guanidinobenzoic acid hydrochloride is a guanidine derivative. Quality standard of 4-guanidinobenzoic acid hydrochloride has been investigated.

Application

4-Guanidinobenzoic acid hydrochloride may be used in ELISA inhibition assay of mouse antiserum. It may be used in the synthesis of human acrosin inhibitor 4′-acetaminophenyl 4-guanidinobenzoate hydrochloride.2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Ramjee et al.
Thrombosis research, 98(6), 559-569 (2000-07-19)
Nafamostat mesilate (FUT-175), a synthetic serine protease inhibitor, is active against a number of the serine proteases involved in coagulation. This has been proposed as the basis of its anticoagulant activity. We investigated the reaction of Nafamostat with bovine pancreatic
I Midgley et al.
Xenobiotica; the fate of foreign compounds in biological systems, 24(1), 79-92 (1994-01-01)
1. The metabolic fate of N,N-dimethylcarbamoylmethyl 4-(4-guanidino[14C]benzoyloxy)phenylacetate methanesulphonate (14C-camostat mesylate) was investigated after i.v. administration to man (12-h infusion), and to rat and dog (bolus injection). 2. Renal excretion (mainly in 24 h) accounted for at least 80% dose in
K Beckh et al.
Research in experimental medicine. Zeitschrift fur die gesamte experimentelle Medizin einschliesslich experimenteller Chirurgie, 187(6), 401-406 (1987-01-01)
The elimination of the low molecular weight proteinase inhibitor camostate (FOY 305) was studied in rats after oral administration and in the the situ perfused rat liver. After feeding of camostate (400 mg/kg b.w.) only the metabolites (FOY 251, GBA)
G T Layton et al.
Clinical and experimental immunology, 69(1), 157-165 (1987-07-01)
Polyclonal and monoclonal antibodies to the antibacterial agent chlorhexidine (1,1'-hexamethylene bis [5-(p-chlorophenyl)]biguanide, mol. wt = 505) were raised using a chlorine-generated N-chloro chlorhexidine-keyhole limpet haemocyanin (NCC-KLH) conjugate as the immunogen. Antibodies were detected by ELISA, using a semi-chlorhexidine derivative conjugated
K Beckh et al.
International journal of pancreatology : official journal of the International Association of Pancreatology, 10(3-4), 197-205 (1991-11-01)
The hepatic metabolism and biliary and pancreatic excretion of the serine protease inhibitor camostat mesilate and its metabolites FOY-251 and GBA were studied in rats in vivo and in in sutu liver-perfusion experiments. After oral feeding (100 mg/kg) and iv

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service