All Photos(1)
About This Item
Linear Formula:
ClCH2C(OCH3)=CHCO2CH3
CAS Number:
Molecular Weight:
164.59
Beilstein:
5376618
MDL number:
UNSPSC Code:
12352103
Assay
97%
refractive index
n20/D 1.484 (lit.)
density
1.21 g/mL at 25 °C (lit.)
SMILES string
COC(=O)\C=C(/CCl)OC
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service