Skip to Content
Merck
All Photos(1)

Documents

357952

Sigma-Aldrich

1-Isoquinolinecarbonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

90-92 °C (lit.)

SMILES string

N#Cc1nccc2ccccc12

InChI

1S/C10H6N2/c11-7-10-9-4-2-1-3-8(9)5-6-12-10/h1-6H

InChI key

HJHXYSBRTVFEDD-UHFFFAOYSA-N

General description

Effect of magnetic field on the photosubstitution reaction of 1-isoquinolinecarbonitrile in ethanol has been reported. Conjugated polynitrile, plasma-polymerized 1-isoquinolinecarbonitrile (PPIQCN) has been prepared by plasma polymerization.

Application

1-Isoquinolinecarbonitrile may be used as starting reagent in the syntheses of imidazo[5,1-a]isoquinolines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization and optical property of novel conjugated polynitrile.
Zhao X-Y, et al.
Plasma Processes and Polymers, 3(4-5), 333-337 (2006)
Kondensierte Isochinoline, XI. Imidazo [5, 1-a] isochinoline.
Reimlinger H, et al.
Chemische Berichte, 108(12), 3771-3779 (1975)
Sharmistha Dutta Choudhury et al.
The journal of physical chemistry. B, 110(17), 8850-8855 (2006-04-28)
Laser flash photolysis and an external magnetic field have been used for the study of the interaction of 4-nitroquinoline-1-oxide (4NQO) with some indole derivatives, amino acids, tyrosine and tryptophan, and model proteins, lysozyme and bovine serum albumin. In an aprotic
Tamara Rodríguez-Cabo et al.
Analytical and bioanalytical chemistry, 407(20), 6159-6170 (2015-06-05)
Time-of-flight accurate mass spectrometry (TOF-MS), following a previous chromatographic (gas or liquid chromatography) separation step, is applied to the identification and structural elucidation of quinoline-like alkaloids in honey. Both electron ionization (EI) MS and positive electrospray (ESI+) MS spectra afforded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service