Skip to Content
Merck
All Photos(1)

Key Documents

348511

Sigma-Aldrich

3-Acetyl-2-oxazolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

110-114 °C/2 mmHg (lit.)

mp

69-71 °C (lit.)

SMILES string

CC(=O)N1CCOC1=O

InChI

1S/C5H7NO3/c1-4(7)6-2-3-9-5(6)8/h2-3H2,1H3

InChI key

COCHMAAUXSQMED-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sánchez-Polo et al.
Water research, 42(15), 4163-4171 (2008-09-03)
The main objective of this study was to analyze the effectiveness of technologies based on the use of ozone and activated carbon for the removal of nitroimidazoles from water, considering them as model of pharmaceutical compounds. A study was undertaken
Temperature dependent 1H NMR study of the substrate complex derived from zinc (II) perchlorate,(R,R)-4, 6-dibenzofurandiyl-2, 2'-bis (4-phenyloxazoline), and 3-acetyl-2-oxazolidinone. Evidence for octahedral structure of the substrate complex.
Kanemasa S, et al.
Tetrahedron Letters, 39(41), 7521-7524 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service