Skip to Content
Merck
All Photos(1)

Key Documents

348376

Sigma-Aldrich

2-(Trifluoromethoxy)phenyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4NCO
CAS Number:
Molecular Weight:
203.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.455 (lit.)

density

1.35 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1ccccc1N=C=O

InChI

1S/C8H4F3NO2/c9-8(10,11)14-7-4-2-1-3-6(7)12-5-13/h1-4H

InChI key

CTMGYQHKKIEXKF-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service