Skip to Content
Merck
All Photos(1)

Documents

338575

Sigma-Aldrich

2-Fluoro-6-methoxyphenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(OCH3)OH
CAS Number:
Molecular Weight:
142.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.52 (lit.)

bp

130-131 °C/36 mmHg (lit.)

density

1.23 g/mL at 25 °C (lit.)

SMILES string

COc1cccc(F)c1O

InChI

1S/C7H7FO2/c1-10-6-4-2-3-5(8)7(6)9/h2-4,9H,1H3

InChI key

YZNHPLVFLRSVHY-UHFFFAOYSA-N

Application

2-Fluoro-6-methoxyphenol was used in the preparation of 5′-halogenated resiniferatoxin analogs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang Su Lim et al.
Bioorganic & medicinal chemistry letters, 21(1), 299-302 (2010-11-30)
A series of 5'-halogenated resiniferatoxin analogs have been investigated in order to examine the effect of halogenation in the A-region on their binding and the functional pattern of agonism/antagonism for rat TRPV1 heterologously expressed in Chinese hamster ovary cells. Halogenation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service