Skip to Content
Merck
All Photos(3)

Key Documents

336572

Sigma-Aldrich

2-Amino-3-methylphenol

96%

Synonym(s):

2-Amino-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)OH
CAS Number:
Molecular Weight:
123.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

Assay

96%

mp

149-152 °C (lit.)

SMILES string

Cc1cccc(O)c1N

InChI

1S/C7H9NO/c1-5-3-2-4-6(9)7(5)8/h2-4,9H,8H2,1H3

InChI key

FEDLEBCVFZMHBP-UHFFFAOYSA-N

General description

2-Amino-3-methylphenol is an aminophenol formed by incubating isopropyl-β-D-thiogalactopyranoside-induced E. coli JS996 strain cells with various nitroaromatic compounds.

Application

2-Amino-3-methylphenol was used in the preparation of Schiff base ligands.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and reactivity of novel cyclometallated complexes derived from [C, N, O] terdentate ligands. Crystal structure of [Pd {2, 3, 4-(MeO) 3 C 6 HC (H)[double bond, length half m-dash] N [2-(O) C 6 H 4]}(PPh 3)].
Fernandez A, et al.
New. J. Chem., 26(4), 398-404 (2002)
Venkateswarlu Kadiyala et al.
Applied and environmental microbiology, 69(11), 6520-6526 (2003-11-07)
The predominant bacterial pathway for nitrobenzene (NB) degradation uses an NB nitroreductase and hydroxylaminobenzene (HAB) mutase to form the ring-fission substrate ortho-aminophenol. We tested the hypothesis that constructed strains might accumulate the aminophenols from nitroacetophenones and other nitroaromatic compounds. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service