Skip to Content
Merck
All Photos(1)

Documents

336130

Sigma-Aldrich

Rubidium acetate

98%

Synonym(s):

Acetic acid rubidium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2Rb
CAS Number:
Molecular Weight:
146.53
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

SMILES string

[Rb+].CC([O-])=O

InChI

1S/C2H4O2.Rb/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

FOGKDYADEBOSPL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service