All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H11N
CAS Number:
Molecular Weight:
181.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
99%
mp
90-92 °C (lit.)
SMILES string
Cn1c2ccccc2c3ccccc13
InChI
1S/C13H11N/c1-14-12-8-4-2-6-10(12)11-7-3-5-9-13(11)14/h2-9H,1H3
InChI key
SDFLTYHTFPTIGX-UHFFFAOYSA-N
General description
9-Methylcarbazole is a nitrogen-containing polycyclic aromatic hydrocarbon. Claisen-Schmidt condensation of 3-formyl-9-methylcarbazole with various amides of 3-aminoacetophenone yields N-{3-[3-(9-methyl-9H-carbazol-3-yl)-acryloyl]-phenyl}-benzamide/amide derivatives.
Application
9-Methylcarbazole was used in facile synthesis of an organic electric conducting nanowire.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
G L Kedderis et al.
Molecular pharmacology, 23(3), 758-760 (1983-05-01)
The source of the oxygen atom in the product of the cytochrome P-450-catalyzed N-demethylation of N-methylcarbazole was determined by mass spectral analysis of the carbinolamine precursor of formaldehyde formed during incubation in oxygen 18-enriched medium. Initial experiments demonstrated that N-(hydroxymethyl)carbazole
W Yang et al.
Xenobiotica; the fate of foreign compounds in biological systems, 23(9), 973-982 (1993-09-01)
1. As previously reported (Yang and Davis 1992), N-methylcarbazole (NMC) is converted to N-hydroxymethylcarbazole (NHMC), and 3-hydroxy-N-hydroxymethylcarbazole (3-OH-NHMC), two relatively stable carbinolamine metabolites by the fungus Cunninghamella echinulata (ATCC 9244). Decomposition of these two carbinolamines yields the corresponding dealkylated metabolites
T Shen et al.
Chemical research in toxicology, 7(2), 231-238 (1994-03-01)
The oxidation rate of NADPH is markedly stimulated during the mechanism-based inactivation of cytochrome P450 2B1 by N-methylcarbazole (NMC) in a reconstituted system consisting of NADPH-cytochrome P450 reductase, cytochrome P450 and phospholipid. The stimulation of NADPH oxidation in this system
B O Ibe et al.
Experimental lung research, 20(3), 207-222 (1994-05-01)
N-methylcarbazole (NMC) is a procarcinogenic component of tobacco smoke particulate matter. It is metabolized by liver microsomes into some hydroxylated metabolites such as the potent mutagen N-hydroxymethylcarbazole (NHMC). Lung metabolism and toxicity of NMC is not known. Since the lung
W Yang et al.
Toxicology letters, 60(3), 307-314 (1992-05-01)
The formation of N-hydroxymethylcarbazole (NHMC), carbazole, 1-hydroxy-N-methylcarbazole, 2-hydroxy-N-methylcarbazole, and 3-hydroxy-N-methylcarbazole as products of mammalian liver microsomal metabolism of N-methylcarbazole (NMC) has been documented by several investigators. In previous studies in our laboratory, the fungus Cunninghamella echinulata (ATCC 9244) produced two
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service