Skip to Content
Merck
All Photos(1)

Documents

308951

Sigma-Aldrich

Ethyl thiocyanate

97%

Synonym(s):

Ethyl rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2SCN
CAS Number:
Molecular Weight:
87.14
Beilstein:
1737620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

145 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

CCSC#N

InChI

1S/C3H5NS/c1-2-5-3-4/h2H2,1H3

InChI key

WFCLYEAZTHWNEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The stomatal response to ethyl thiocyanate (ESCN) in Arabidopsis was studied.

Application

Ethyl thiocyanate was used in the synthesis of hydrogen cyanide by undergoing in situ reduction with dithiothreitol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z L Cardeal et al.
Journal of analytical toxicology, 19(1), 31-34 (1995-01-01)
This work compares two different methods for assaying hydrogen cyanide, a spectrophotometric method and a headspace gas chromatographic method, each with its own reference standard generation. In the first method, the reference standards are cyanide solutions. In the second method
Mohammad Shakhawat Hossain et al.
Bioscience, biotechnology, and biochemistry, 77(5), 977-983 (2013-05-08)
Isothiocyanates, nitriles, and thiocyanates are degradation products of glucosinolates in crucifer plants. In this study, we investigated the stomatal response to allyl isothiocyanate (AITC), 3-butenenitrile (3BN), and ethyl thiocyanate (ESCN) in Arabidopsis. AITC, 3BN, and ESCN induced stomatal closure in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service