Skip to Content
Merck
All Photos(1)

Documents

305537

Sigma-Aldrich

Benzyl methyl malonate

90%

Synonym(s):

Methyl benzyl malonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCOCH2COOCH3
CAS Number:
Molecular Weight:
208.21
Beilstein:
2529986
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

90%

form

liquid

impurities

<10% dimethyl malonate

refractive index

n20/D 1.502 (lit.)

bp

125 °C/0.5 mmHg (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CC(=O)OCc1ccccc1

InChI

1S/C11H12O4/c1-14-10(12)7-11(13)15-8-9-5-3-2-4-6-9/h2-6H,7-8H2,1H3

InChI key

IAUZDBFOEWAQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl methyl malonate is an ester.

Application

Benzyl methyl malonate may be used in chemical synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service