Skip to Content
Merck
All Photos(2)

Key Documents

29511

Sigma-Aldrich

(S)-(−)-(6,6′-Dimethoxybiphenyl-2,2′-diyl)bis(diphenylphosphine)

greener alternative

≥97%, optical purity ee: ≥99%

Synonym(s):

(S)-(−)-2,2′-Bis(diphenylphosphino)-6,6′-dimethoxy-1,1′-biphenyl, (S)-(−)-MeO-BIPHEP, SL-A101-2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H32O2P2
CAS Number:
Molecular Weight:
582.61
Beilstein:
4790083
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22
To inquire about this product 29511, please contact your local Merck office or dealer. Contact Technical Service

Quality Level

Assay

≥97%

optical purity

ee: ≥99%

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

functional group

phosphine

greener alternative category

SMILES string

COc1cccc(P(c2ccccc2)c3ccccc3)c1-c4c(OC)cccc4P(c5ccccc5)c6ccccc6

InChI

1S/C38H32O2P2/c1-39-33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)38-34(40-2)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32/h3-28H,1-2H3

InChI key

KRJVQCZJJSUHHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Atropisomeric MeOBIPHEP ligands

  • C–C coupling reactions
  • Ligand for palladium-catalyzed intramolecular allylic alkylation of unsaturated amides
  • Enantioselective hydrogenation of a-amino-ß-keto ester hydrochlorides catalyzed by an iridium complex
  • Asymmetric hydrogenation of aromatic ketones
Ligand for ruthenium-catalyzed greener amine synthesis by enamine reduction with hydrogen gas.

Asymmetric Synthesis of (S)-3-Amino-4-methoxy-butan-1-ol by Way of Reductive Amination

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service