Skip to Content
Merck
All Photos(1)

Documents

294616

Sigma-Aldrich

(+)-Pseudoephedrine hydrochloride

99%

Synonym(s):

(+)-ψ-Ephedrine hydrochloride, (1S,2S)-2-Methylamino-1-phenylpropanol hydrochloride, d-Isoephedrine hydrochloride, d-Pseudoephedrine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH·HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
3915112
EC Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Assay

99%

optical activity

[α]25/D +61°, c = 1 in H2O

mp

185-188 °C (lit.)

SMILES string

Cl.CN[C@@H](C)[C@@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10+;/m0./s1

InChI key

BALXUFOVQVENIU-KXNXZCPBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-selective adrenergic agonist; decongestant

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(12), 2302-2308 (2010-09-17)
Drug-induced liver injury (DILI) is one of the most important reasons for drug development failure at both preapproval and postapproval stages. There has been increased interest in developing predictive in vivo, in vitro, and in silico models to identify compounds
Zhichao Liu et al.
PLoS computational biology, 7(12), e1002310-e1002310 (2011-12-24)
Drug-induced liver injury (DILI) is a significant concern in drug development due to the poor concordance between preclinical and clinical findings of liver toxicity. We hypothesized that the DILI types (hepatotoxic side effects) seen in the clinic can be translated
Nigel Greene et al.
Chemical research in toxicology, 23(7), 1215-1222 (2010-06-18)
Drug-induced liver injury is a major issue of concern and has led to the withdrawal of a significant number of marketed drugs. An understanding of structure-activity relationships (SARs) of chemicals can make a significant contribution to the identification of potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service