Skip to Content
Merck
All Photos(1)

Documents

284378

Sigma-Aldrich

1,1′-Diethyl-2,4′-cyanine iodide

Dye content 97 %

Synonym(s):

Isocyanine iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H23IN2
CAS Number:
Molecular Weight:
454.35
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, 97%

mp

158-160 °C (lit.)

λmax

558 nm

SMILES string

[I-].CCN1\C(C=Cc2ccccc12)=C\c3cc[n+](CC)c4ccccc34

InChI

1S/C23H23N2.HI/c1-3-24-16-15-19(21-10-6-8-12-23(21)24)17-20-14-13-18-9-5-7-11-22(18)25(20)4-2;/h5-17H,3-4H2,1-2H3;1H/q+1;/p-1

InChI key

MYKATFBGLTVMQX-UHFFFAOYSA-M

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service