Recommended Products
Assay
97%
form
liquid
reaction suitability
reagent type: reductant
refractive index
n20/D 1.376 (lit.)
bp
105-107 °C/751 mmHg (lit.)
density
0.781 g/mL at 25 °C (lit.)
SMILES string
CB(OC(C)C)OC(C)C
InChI
1S/C7H17BO2/c1-6(2)9-8(5)10-7(3)4/h6-7H,1-5H3
InChI key
BHOFOFKBMGVFSG-UHFFFAOYSA-N
Application
Potential derivatizing agent for difunctional compounds for GC and GC-MS analysis.
Reactant for:
- Preparation of alkylborate compounds containing imidazolyl functional groups
- Nickel complexes of monoanionic tripodal ligands (carboxylate shift from nickel to boron)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
44.6 °F - closed cup
Flash Point(C)
7 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service