Skip to Content
Merck
All Photos(2)

Key Documents

279617

Sigma-Aldrich

2-Phenyl-1,3-dithiane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12S2
CAS Number:
Molecular Weight:
196.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

72-74 °C (lit.)

solubility

acetone: soluble 25 mg/mL, clear, colorless to yellow

functional group

phenyl
thioether

SMILES string

C1CSC(SC1)c2ccccc2

InChI

1S/C10H12S2/c1-2-5-9(6-3-1)10-11-7-4-8-12-10/h1-3,5-6,10H,4,7-8H2

InChI key

GXKPARDRBFURON-UHFFFAOYSA-N

General description

The full set of NMR spectral parameters (1H, 13C, 31P) for 2-phenyl-1,3-dithiane has been reported.

Application

2-Phenyl-1,3-dithiane is a versatile acyl anion equivalent and has been used in the preparation of benzaldehyde-1-d.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 1-deuterioaldehydes. Benzaldehyde-1-D.
Seebach D, et al.
The Journal of Organic Chemistry, 31(12), 4303-4304 (1966)
NMR spectral analysis of 1, 3, 2-dithiaphosphorinanes and 1, 3-dithianes.
Martin J and Robert JB.
Org. Magn. Reson., 7(2), 76-77 (1975)
Smith, A. B., III; Boldi, A. M.
Journal of the American Chemical Society, 119, 6925-6925 (1997)
Amos B Smith et al.
Journal of the American Chemical Society, 125(47), 14435-14445 (2003-11-20)
The development, application, and advantages of a one-flask multicomponent dithiane linchpin coupling protocol, over the more conventional stepwise addition of dithiane anions to electrophiles leading to the rapid, efficient, and stereocontrolled assembly of highly functionalized intermediates for complex molecule synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service