Skip to Content
Merck
All Photos(1)

Key Documents

275131

Sigma-Aldrich

Bis(1,5-cyclooctadiene)diiridium(I) dichloride

97%

Synonym(s):

[Ir(1,5-cod)Cl]2, 1,5-Cyclooctadiene-iridium(I) chloride dimer, Chloro(1,5-cyclooctadiene)iridium(I) dimer, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]diiridium, Iridium(I) chloride 1,5-cyclooctadiene complex dimer, [Ir(1,5-cod)Cl]2, [Ir(cod)Cl]2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24Cl2Ir2
CAS Number:
Molecular Weight:
671.70
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

reagent type: catalyst
core: iridium

mp

205 °C (dec.) (lit.)

SMILES string

Cl[Ir].Cl[Ir].C1CC=CCCC=C1.C2CC=CCCC=C2

InChI

1S/2C8H12.2ClH.2Ir/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1-2,7-8H,3-6H2;2*1H;;/q;;;;2*+1/p-2/b2*2-1-,8-7-;;;;

InChI key

ZFOUDQNHNLDNLD-MIXQCLKLSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Metal precursor for asymmetric allylic substitutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P A Marks et al.
Journal of the National Cancer Institute, 92(15), 1210-1216 (2000-08-03)
Histone deacetylase (HDAC) inhibitors have been shown to be potent inducers of growth arrest, differentiation, and/or apoptotic cell death of transformed cells in vitro and in vivo. One class of HDAC inhibitors, hydroxamic acid-based hybrid polar compounds (HPCs), induce differentiation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service