Skip to Content
Merck
All Photos(2)

Documents

269018

Sigma-Aldrich

2-Phenyl-2-butanol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(C2H5)(CH3)OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.519 (lit.)

bp

107-108 °C/20 mmHg (lit.)

density

0.977 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(O)c1ccccc1

InChI

1S/C10H14O/c1-3-10(2,11)9-7-5-4-6-8-9/h4-8,11H,3H2,1-2H3

InChI key

XGLHYBVJPSZXIF-UHFFFAOYSA-N

Application

2-Phenyl-2-butanol was used in the synthesis of 2-phenylbutane in trifluoroacetic acid-dichloromethane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric reductions of carbocations by chiral organosilicon hydrides. The stereochemical nature of the carbocation captured.
Journal of the American Chemical Society, 100(4), 7641-7644 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service