All Photos(1)
About This Item
Linear Formula:
O2NC6H(OCH3)3CO2CH3
CAS Number:
Molecular Weight:
271.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
mp
67-69 °C (lit.)
SMILES string
COC(=O)c1cc(OC)c(OC)c(OC)c1[N+]([O-])=O
InChI
1S/C11H13NO7/c1-16-7-5-6(11(13)19-4)8(12(14)15)10(18-3)9(7)17-2/h5H,1-4H3
InChI key
HCEIEWYPDCDNNU-UHFFFAOYSA-N
Application
Methyl 2-nitro-3,4,5-trimethoxybenzoate has been used in preparation of 2-nitro-3,4,5-trimethoxybenzoic acid.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Gang Liu et al.
Bioorganic & medicinal chemistry, 15(20), 6608-6617 (2007-08-08)
A series of 4-aminoquinazoline derivatives is prepared by the nucleophilic substitution reaction of 6,7,8-trimethoxy-4-chloroquinazoline and aryl amine. The structures of the compounds are confirmed by elemental analysis, IR, and (1)H NMR spectral data. The compounds are also evaluated for their
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service