Skip to Content
Merck
All Photos(1)

Documents

247081

Sigma-Aldrich

3,5-Bis(trifluoromethyl)phenol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3OH
CAS Number:
Molecular Weight:
230.11
Beilstein:
2123670
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.415 (lit.)

bp

97 °C/50 mmHg (lit.)

mp

20-21 °C (lit.)

density

1.511 g/mL at 25 °C (lit.)

SMILES string

Oc1cc(cc(c1)C(F)(F)F)C(F)(F)F

InChI

1S/C8H4F6O/c9-7(10,11)4-1-5(8(12,13)14)3-6(15)2-4/h1-3,15H

InChI key

ODSXJQYJADZFJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Bis(trifluoromethyl)phenol (BTMP) has been used as hydrogen donor in synergistic extraction of aluminium (III) and gallium (III) with 2,4-pentanedione in heptane.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steric effect of 3, 5-bis (trifluoromethyl) phenol as hydrogen-bond donor on outer-sphere complexation in the synergistic extraction of aluminium (III) and gallium (III) with 2, 4-pentanedione.
Imura H, et al.
Bulletin of the Chemical Society of Japan, 78(12), 2146-2151 (2005)
Douglas H Read et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 35(6), 671-677 (2019-02-19)
We describe for the first time hydrogen bonded acid (HBA) polymer, poly{methyl[3-(2-hydroxyl, 4,6-bistrifluoromethyl)phenyl]propylsiloxane}, (DKAP), as stationary phase for gas chromatography (μGC) of organophosphate (OP), chemical warfare agent (CWA) surrogates, dimethylmethylphosphonate (DMMP), diisopropylmethylphosphonate (DIMP), diethylmethylphosphonate (DEMP), and trimethylphosphate (TMP), with high
Nobuyuki Kakutani et al.
Xenobiotica; the fate of foreign compounds in biological systems, 51(1), 88-94 (2020-09-03)
The formation of reactive metabolites (RMs) is a problem in drug development that sometimes results in severe hepatotoxicity. As detecting RMs themselves is difficult, a covalent binding assay using expensive radiolabelled tracers is usually performed for candidate selection. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service