Skip to Content
Merck
All Photos(1)

Key Documents

238694

Sigma-Aldrich

IGEPAL® DM-970

Synonym(s):

Polyoxyethylene (150) dinonylphenyl ether, Polyoxyethylene, dinonylphenyl and nonylphenyl ethers, branched

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n · C24H42O · C15H24O
MDL number:
UNSPSC Code:
12162002

mp

59-61 °C (lit.)

HLB

19

Legal Information

IGEPAL is a registered trademark of Solvay

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>500.0 °F - closed cup

Flash Point(C)

> 260 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

EU REACH Annex XIV (Authorisation List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service